Learn More
Waters Corp Waters Tubing Assembly, System Outlet, 2535Q
Catalog No. 50791967 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for connecting the filter outlet port to the injector inlet port. It is a critical component for fluidic pathways in specific systems.
  • Used for connecting the 25X5QGM filter outlet port to the injector inlet port.
  • Functions as a system outlet tube.
  • Included as a component in a larger startup kit.
Catalog No. 50-791-967 Supplier Waters Corp Supplier No. 430002273
This tubing assembly is designed for connecting the filter outlet port to the injector inlet port. It is a critical component for fluidic pathways in specific systems.
  • Used for connecting the 25X5QGM filter outlet port to the injector inlet port.
  • Functions as a system outlet tube.
  • Included as a component in a larger startup kit.