Learn More
Waters Corp Waters Corporation Tubing Assembly, System Outlet, 24 in., 0.005 in. ID, Stainless Steel
Catalog No. 50466549 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement outlet tube, constructed from stainless steel. It features a 24 in. length and a 0.005 in. internal diameter, designed for connecting an ACQUITY Binary Solvent Manager to a Sample Manager.
  • 24 in. length
  • 0.005 in. internal diameter
  • Stainless steel construction
  • Connects ACQUITY Binary Solvent Manager to Sample Manager
Catalog No. 50-466-549 Supplier Waters Corp Supplier No. 430001017
This tubing assembly is a replacement outlet tube, constructed from stainless steel. It features a 24 in. length and a 0.005 in. internal diameter, designed for connecting an ACQUITY Binary Solvent Manager to a Sample Manager.
  • 24 in. length
  • 0.005 in. internal diameter
  • Stainless steel construction
  • Connects ACQUITY Binary Solvent Manager to Sample Manager