Learn More
Waters Corp Waters Tubing Assembly, Transducer to Check Valve, Stainless Steel, UPLC
Catalog No. 50791937 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tube assembly is designed to connect the transducer to the check valve, serving as a critical component of the Intelligent Intake Valve found within the ACQUITY Binary Solvent Manager.
  • Made from stainless steel for durability and chemical compatibility.
  • Designed for UPLC system types.
  • Provides a secure connection between the transducer and the check valve.
  • Essential component for the Intelligent Intake Valve.
Catalog No. 50-791-937 Supplier Waters Corp Supplier No. 430001773
This tube assembly is designed to connect the transducer to the check valve, serving as a critical component of the Intelligent Intake Valve found within the ACQUITY Binary Solvent Manager.
  • Made from stainless steel for durability and chemical compatibility.
  • Designed for UPLC system types.
  • Provides a secure connection between the transducer and the check valve.
  • Essential component for the Intelligent Intake Valve.