Learn More
Waters Corp WATERS CORPORATION Tubing Assembly, Transducer to Vent Valve, UPLC System
Catalog No. 50791968 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly connects the transducer to the vent valve for specific high-performance liquid chromatography systems.
  • Designed for use with ISM and QSM system components.
  • Compatible with H-Class system and APC instrumentation.
  • Each package contains one unit.
  • This item is a tubing component.
  • Replacement should be performed as needed.
Catalog No. 50-791-968 Supplier Waters Corp Supplier No. 430002316
This tubing assembly connects the transducer to the vent valve for specific high-performance liquid chromatography systems.
  • Designed for use with ISM and QSM system components.
  • Compatible with H-Class system and APC instrumentation.
  • Each package contains one unit.
  • This item is a tubing component.
  • Replacement should be performed as needed.