Learn More
Med Vet International BD Vacutainer SST™ Venous Blood Collection Tube Clot Activator / Separator Gel Additive 8.5 mL Conventional Closure Plastic Tube, 367988, 1000/Case
Catalog No. 503043407
Encompass_Preferred
Clot activator and gel for serum separation
Catalog No. 50-304-3407 Supplier Med Vet International Supplier No. 207067CS
Clot activator and gel for serum separation