Learn More
Med Vet International VACUETTE Z Serum Sep Clot Activator Venous Blood Collection Tube Clot Activator / Separator Gel Additive 5 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 456073P, 50/Box
Catalog No. 503044847
Encompass_Preferred
Greiner's Vacuette CAT serum separator clot activator tubes contain a separating gel that migrates up to the interface between serum and clot during centrifugation, where it forms a stable barrier and separates the serum from the fibrin and cells Sterile, 5 mL serum tube has a red pull cap with a yellow ring Dimensions: 13 x 100 millimeters Made of clear, polyethylene terephthalate (PET)
Catalog No. 50-304-4847 Supplier Med Vet International Supplier No. 416968BX
Greiner's Vacuette CAT serum separator clot activator tubes contain a separating gel that migrates up to the interface between serum and clot during centrifugation, where it forms a stable barrier and separates the serum from the fibrin and cells Sterile, 5 mL serum tube has a red pull cap with a yellow ring Dimensions: 13 x 100 millimeters Made of clear, polyethylene terephthalate (PET)