Learn More
Med Vet International VACUETTE Z Serum Sep Clot Activator Venous Blood Collection Tube Clot Activator / Separator Gel Additive 8 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 455071P, 50/Box
Catalog No. 503042857
Encompass_Preferred
Greiner's Vacuette CAT serum separator clot activator tubes contain a separating gel that migrates up to the interface between serum and clot during centrifugation, where it forms a stable barrier and separates the serum from the fibrin and cells 8 mL serum tubes come with an additive that serves as a clot activator and separator gel Pull cap is color coded red with a yellow ring Dimensions: 16 x 100 mm Clear, silica-coated tube is made of polyethylene terephthalate (PET)
Catalog No. 50-304-2857 Supplier Med Vet International Supplier No. 409479BX
Greiner's Vacuette CAT serum separator clot activator tubes contain a separating gel that migrates up to the interface between serum and clot during centrifugation, where it forms a stable barrier and separates the serum from the fibrin and cells 8 mL serum tubes come with an additive that serves as a clot activator and separator gel Pull cap is color coded red with a yellow ring Dimensions: 16 x 100 mm Clear, silica-coated tube is made of polyethylene terephthalate (PET)