Learn More
Med Vet International VACUETTE Venous Blood Collection Tube K2 EDTA Additive 4 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 454209, 50/Pack
Catalog No. 503044378
Encompass_Preferred
The interior of these Vacuette tubes is coated with K2 EDTA – an anticoagulant that prevents blood from clotting by inhibiting the action of clotting factors Pull caps are lavender with black rings Dimensions:13 x 75 millimeters Volume: 3 mL Clear, silica-coated tube is polyethylene terephthalate (PET)
Catalog No. 50-304-4378 Supplier Med Vet International Supplier No. 505509PK
The interior of these Vacuette tubes is coated with K2 EDTA – an anticoagulant that prevents blood from clotting by inhibiting the action of clotting factors Pull caps are lavender with black rings Dimensions:13 x 75 millimeters Volume: 3 mL Clear, silica-coated tube is polyethylene terephthalate (PET)