Learn More
Ace Glass, Inc. TUBE BLEED 10/18 TUBE BLEED 10/18
SDP
Catalog No. 50976728
Encompass_Preferred
Bleed tube with 10/18 joint. Used with short path distillation apparatus designed for small quantities where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with the accuracy of larger setups.
Catalog No. 50-976-728 Supplier Ace Glass, Inc. Supplier No. 931717
Bleed tube with 10/18 joint. Used with short path distillation apparatus designed for small quantities where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with the accuracy of larger setups.