Learn More
SKC Inc SKC Tube Breaker
SDP
Catalog No. NC1294355
Encompass
All-in-One SKC Tube Breaker opens glass sorbent tubes cleanly and safely. Stainless steel, can be used with 6,7,8, and 10-mm OD tubes. Heavy-duty cap rotates to expose the desired size opener and to retain glass tips to be discarded. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC1294355 Supplier SKC Inc Supplier No. 22603055
All-in-One SKC Tube Breaker opens glass sorbent tubes cleanly and safely. Stainless steel, can be used with 6,7,8, and 10-mm OD tubes. Heavy-duty cap rotates to expose the desired size opener and to retain glass tips to be discarded. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”