Learn More
MSC Vikan Tube Brush, 0.8 in, 19.7 in, Medium, Yellow
Catalog No. 502836555
Encompass
This Vikan tube brush, 0.8 in diameter and 19.7 in length, effectively cleans bottles, tubes, and hard-to-reach surfaces. Constructed with durable polyester bristles, a polypropylene handle, and a stainless steel shaft, this food-safe brush features medium stiffness for efficient cleaning in demanding environments.
  • Cleans bottles, tubes, and hard-to-reach surfaces.
  • Durable polyester bristles, polypropylene handle, and stainless steel shaft.
  • Medium bristle stiffness removes dirt and residue.
  • Yellow color supports HACCP color-coded systems.
  • Complies with EC 1935/2004, FDA CFR 21, UK 2019 No. 704 for food contact.
  • Dishwasher and autoclave safe up to 249.8 °F.
  • BPA free.
  • Ergonomic handle for comfortable grip.
Catalog No. 50-283-6555 Supplier MSC Supplier No. 50264464
This Vikan tube brush, 0.8 in diameter and 19.7 in length, effectively cleans bottles, tubes, and hard-to-reach surfaces. Constructed with durable polyester bristles, a polypropylene handle, and a stainless steel shaft, this food-safe brush features medium stiffness for efficient cleaning in demanding environments.
  • Cleans bottles, tubes, and hard-to-reach surfaces.
  • Durable polyester bristles, polypropylene handle, and stainless steel shaft.
  • Medium bristle stiffness removes dirt and residue.
  • Yellow color supports HACCP color-coded systems.
  • Complies with EC 1935/2004, FDA CFR 21, UK 2019 No. 704 for food contact.
  • Dishwasher and autoclave safe up to 249.8 °F.
  • BPA free.
  • Ergonomic handle for comfortable grip.