Learn More
Ace Glass, Inc. TUBE C TUBE C
SDP
Supplier:  Ace Glass, Inc. 669632
Encompass_Preferred
50mL freeze dry and storage tube with PTFE lined screw cap and stops on the side to prevent ¿suck in¿ under vacuum. Tube fits a #25 Ace ¿Thred. Working capacity of tube is about ¿ of volume. 25mm O.D. x 150mm L.
Catalog No. 50-973-324