Learn More
Covaris Inc Tube & Cap | 16x100mm
SDP
Catalog No. NC1812850
Encompass
This tube and cap combo has a teflon seal and sample volume of 12ml. The suitable application for this item is biomarker extraction.
Catalog No. NC1812850 Supplier Covaris Inc Supplier No. 520191
This tube and cap combo has a teflon seal and sample volume of 12ml. The suitable application for this item is biomarker extraction.