Learn More
Cole-Parmer Argos Technologies Flip-Cap Centrifuge Tubes, 15 mL; 25/Bag, 20 Bags/Cs
Catalog No. 501960590 Shop All Cole-Parmer Products
Encompass_Preferred
Centrifuge tubes with hinged flip-top caps allow for easy one-handed opening and closing. No more dropping or losing the tube cap. Bold black graduations are easy to read and accurate to ±2%. A large frosted marking area allows for permanent sample identification.
The conical-bottom, polypropylene (PP) tubes are compatible with any fixed angle or swing bucket rotor and can withstand temperatures from -80 to 120°C.
Catalog No. 50-196-0590 Supplier Cole-Parmer Supplier No. 0439555
Centrifuge tubes with hinged flip-top caps allow for easy one-handed opening and closing. No more dropping or losing the tube cap. Bold black graduations are easy to read and accurate to ±2%. A large frosted marking area allows for permanent sample identification.
The conical-bottom, polypropylene (PP) tubes are compatible with any fixed angle or swing bucket rotor and can withstand temperatures from -80 to 120°C.