Learn More
Wilmad Labglass TUBE CENT OIL ASTM 100ML
Catalog No. 50806304
Encompass
Tube, Centrifuge, Oil, ASTM. Capacity: 100mL. Calibrated to contain. Used for the determination of water and sediment in petroleum and its products. See ASTM methods D 96, D 1796, and D91.
Catalog No. 50-806-304 Supplier Wilmad Labglass Supplier No. LG4470100
Tube, Centrifuge, Oil, ASTM. Capacity: 100mL. Calibrated to contain. Used for the determination of water and sediment in petroleum and its products. See ASTM methods D 96, D 1796, and D91.