Learn More
SETON SCIENTIFIC TUBE CENT POL1X3.5 38.5ML 5OPK
Catalog No. NC9115402
Encompass
Tube, Open-Top Polyclear (Ultra); 1 x 3-1/2 in., 25 x 89mm; 38.5mL Capacity Tube; Used in the following rotors Beckman Rotor SW32, SW28, SW27, 70Ti, 60Ti, 55.2Ti, 50.2Ti, 42.1
Catalog No. NC9115402 Supplier SETON SCIENTIFIC Supplier No. 7052
Tube, Open-Top Polyclear (Ultra); 1 x 3-1/2 in., 25 x 89mm; 38.5mL Capacity Tube; Used in the following rotors Beckman Rotor SW32, SW28, SW27, 70Ti, 60Ti, 55.2Ti, 50.2Ti, 42.1