Learn More
Med Vet International McKesson Centrifuge Tube, 15 mL, 177-6285, 500/Case
Catalog No. 503042898
Encompass_Preferred
McKesson Centrifuge Tube Conical Bottom Plain 17 X 120 mm 15 mL Blue Screw Cap Polypropylene Tube 15 mL centrifuge tube 17 x 120 millimeters High-quality, translucent tubes molded from polypropylene material featuring a conical bottom RCF rating: 8,078xG Black printed graduations in 0.5 mL increments for easy measurements Large, white printed writing space for convenient labeling Sterile tubes are autoclavable (without cap) for continued use Blue screw caps made of high density polyethylene (HDPE) are flat-top and leak-resistant Racked with color-coded screw caps attached for added convenience
Catalog No. 50-304-2898 Supplier Med Vet International Supplier No. 1082079CS
McKesson Centrifuge Tube Conical Bottom Plain 17 X 120 mm 15 mL Blue Screw Cap Polypropylene Tube 15 mL centrifuge tube 17 x 120 millimeters High-quality, translucent tubes molded from polypropylene material featuring a conical bottom RCF rating: 8,078xG Black printed graduations in 0.5 mL increments for easy measurements Large, white printed writing space for convenient labeling Sterile tubes are autoclavable (without cap) for continued use Blue screw caps made of high density polyethylene (HDPE) are flat-top and leak-resistant Racked with color-coded screw caps attached for added convenience