Learn More
Chemglass Life Sciences Chemglass 250 mL Conical Centrifuge Tube, Polypropylene with HDPE Cap - Racked (6/rack, 24/case)
SDP
Catalog No. 502733409
Encompass_Preferred
Chemglass 250 mL conical centrifuge tubes are designed for high-volume sample processing and efficient pelleting in life science and clinical laboratories. The clear, graduated tubes provide reliable volume measurement and are supplied racked for convenient handling and automation.
  • Polypropylene conical tube with high-density polyethylene cap.
  • 250 mL capacity with clear graduated scale for accurate measurement.
  • Electron beam sterilized; DNase-, RNase-, and pyrogen-free.
  • Racked packaging for efficient handling: 6 per rack, 24 per case.
  • Suitable for separation and harvest of large cells, supernatants, bacteria, yeast, and tissue samples.
Catalog No. 50-273-3409 Supplier Chemglass Life Sciences Supplier No. CGN4306R250
Chemglass 250 mL conical centrifuge tubes are designed for high-volume sample processing and efficient pelleting in life science and clinical laboratories. The clear, graduated tubes provide reliable volume measurement and are supplied racked for convenient handling and automation.
  • Polypropylene conical tube with high-density polyethylene cap.
  • 250 mL capacity with clear graduated scale for accurate measurement.
  • Electron beam sterilized; DNase-, RNase-, and pyrogen-free.
  • Racked packaging for efficient handling: 6 per rack, 24 per case.
  • Suitable for separation and harvest of large cells, supernatants, bacteria, yeast, and tissue samples.