Learn More
Chemglass Life Sciences Chemglass Centrifuge Tube, Conical Bottom, 500 mL, Polypropylene (CGN-4306-R500)
SDP
Catalog No. 502733408
Encompass_Preferred
Chemglass 500 mL conical centrifuge tube molded from clear polypropylene with a screw cap designed for secure sealing. Available in racked and bulk packaging, the tube is intended for separation and harvest of biological and chemical samples and is suitable for high-throughput laboratory workflows.
  • 500 mL nominal volume.
  • Clear polypropylene construction for chemical compatibility and visibility.
  • High-density polyethylene cap with double-seal design for improved leak resistance.
  • Electron beam sterilized with a sterility assurance level (SAL) of 10^-6.
  • DNase-free, RNase-free, and pyrogen-free.
  • Conical bottom with increased thickness for higher centrifugal limits.
  • Packaging options: racked (4 pcs/rack, 24 pcs/case) and bulk (6 pcs/pack, 36 pcs/case).
Catalog No. 50-273-3408 Supplier Chemglass Life Sciences Supplier No. CGN4306R500
Chemglass 500 mL conical centrifuge tube molded from clear polypropylene with a screw cap designed for secure sealing. Available in racked and bulk packaging, the tube is intended for separation and harvest of biological and chemical samples and is suitable for high-throughput laboratory workflows.
  • 500 mL nominal volume.
  • Clear polypropylene construction for chemical compatibility and visibility.
  • High-density polyethylene cap with double-seal design for improved leak resistance.
  • Electron beam sterilized with a sterility assurance level (SAL) of 10^-6.
  • DNase-free, RNase-free, and pyrogen-free.
  • Conical bottom with increased thickness for higher centrifugal limits.
  • Packaging options: racked (4 pcs/rack, 24 pcs/case) and bulk (6 pcs/pack, 36 pcs/case).