Learn More
Chemglass Life Sciences Tube, Centrifuge, 500mL, Conical Bottom, Bulk Packed: 6pcs/Pack, 36pcs/Case
SDP
Catalog No. 5000980728
Encompass_Preferred
The 250 and 500mL centrifuge tubes can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences, clinical experiments and can meet the requirements of biological analysis. Unique structure design of the cap provides double security, further improving the tight seal ability. Increased the thickness of conical bottom in order to improve the maximum centrifugal limit. Polypropylene tube and high-density polyethylene cap. Max. centrifugal limit 7000xg. Temperature range: -80°C ~120°C. Clear grad scale. DNase, RNase and pyrogen free. Electron beam sterilization, SAL=10-6. Separation of sediments after chemical reactions for harvest and application. Harvest of biomacromolecules, inorganics and organics.
Catalog No. 50-009-80728 Supplier Chemglass Life Sciences Supplier No. CGN4306500
The 250 and 500mL centrifuge tubes can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences, clinical experiments and can meet the requirements of biological analysis. Unique structure design of the cap provides double security, further improving the tight seal ability. Increased the thickness of conical bottom in order to improve the maximum centrifugal limit. Polypropylene tube and high-density polyethylene cap. Max. centrifugal limit 7000xg. Temperature range: -80°C ~120°C. Clear grad scale. DNase, RNase and pyrogen free. Electron beam sterilization, SAL=10-6. Separation of sediments after chemical reactions for harvest and application. Harvest of biomacromolecules, inorganics and organics.