Learn More
Med Vet International McKesson Conical Bottom Centrifuge Tube, 30 x 115 mm, 177-6288, 25/Bag
Catalog No. 503043829
Encompass_Preferred
McKesson Centrifuge Tube Conical Bottom Plain 30 X 115 mm 50 mL Blue Screw Cap Polypropylene Tube Volume of tubes is 50 mL Black printed graduations in 0.5 mL increments for easy measurements Large white printed writing space for convenient labeling Autoclavable tubes for continued use (without cap) Flat-top, leak-resistant, high-density polyethylene (HDPE) screw caps are included
Catalog No. 50-304-3829 Supplier Med Vet International Supplier No. 1082082BG
 Restricted Item
Add to Cart
Add to Cart
McKesson Centrifuge Tube Conical Bottom Plain 30 X 115 mm 50 mL Blue Screw Cap Polypropylene Tube Volume of tubes is 50 mL Black printed graduations in 0.5 mL increments for easy measurements Large white printed writing space for convenient labeling Autoclavable tubes for continued use (without cap) Flat-top, leak-resistant, high-density polyethylene (HDPE) screw caps are included