Learn More
Med Vet International McKesson Conical Bottom Centrifuge Tube, 30 x 115 mm, 177-6289, 500/Case
Catalog No. 503042904
Encompass_Preferred
McKesson Centrifuge Tube Conical Bottom Plain 30 X 115 mm 50 mL Blue Screw Cap Polypropylene Tube 50 mL translucent polypropylene centrifuge tube with conical bottom 30 x 115 millimeters 8,078xG / 8500 RCF Spin Rate RCF rating: 8,078xG Black printed graduations in 0.5 mL increments for easy measurements Large, white printed writing space for convenient labeling Sterile tubes are autoclavable (without cap) for continued use Blue screw caps made of high density polyethylene (HDPE) are flat-top and leak-resistant Racked with color-coded screw caps attached for added convenience
Catalog No. 50-304-2904 Supplier Med Vet International Supplier No. 1082083CS
McKesson Centrifuge Tube Conical Bottom Plain 30 X 115 mm 50 mL Blue Screw Cap Polypropylene Tube 50 mL translucent polypropylene centrifuge tube with conical bottom 30 x 115 millimeters 8,078xG / 8500 RCF Spin Rate RCF rating: 8,078xG Black printed graduations in 0.5 mL increments for easy measurements Large, white printed writing space for convenient labeling Sterile tubes are autoclavable (without cap) for continued use Blue screw caps made of high density polyethylene (HDPE) are flat-top and leak-resistant Racked with color-coded screw caps attached for added convenience