Learn More
Thomas Scientific 50mL Centrifuge Tube, PS Rack, Sterile
SDP
Catalog No. 501457572
Encompass
Printed Graduations (with marking area) Polypropolyne for the tube and polyethylene for the cap High centrifugal strength - up to 10,000 x g Flat markable cap with leakproof seal One-hand operation design for the caps provides easy handling Sterilized by E-Beam DNase, RNase and pyrogen free
Catalog No. 50-145-7572 Supplier Thomas Scientific Supplier No. 1194Z49
Printed Graduations (with marking area) Polypropolyne for the tube and polyethylene for the cap High centrifugal strength - up to 10,000 x g Flat markable cap with leakproof seal One-hand operation design for the caps provides easy handling Sterilized by E-Beam DNase, RNase and pyrogen free