Learn More
Med Vet International McKesson Centrifuge Tube, 12 mL, 177-112010, 500/Bag
Catalog No. 503044026
Encompass_Preferred
McKesson Centrifuge Tube Conical Bottom Plain 16 X 100 mm 12 mL Without Color Coding Without Closure Polystyrene Tube Nonsterile, conical bottom polystyrene centrifuge tubes are without closure or color coding Dimensions are 16 x 100 mm Volume is 12 mL RCF rating is 1,500xG
Catalog No. 50-304-4026 Supplier Med Vet International Supplier No. 1175302BG
McKesson Centrifuge Tube Conical Bottom Plain 16 X 100 mm 12 mL Without Color Coding Without Closure Polystyrene Tube Nonsterile, conical bottom polystyrene centrifuge tubes are without closure or color coding Dimensions are 16 x 100 mm Volume is 12 mL RCF rating is 1,500xG