Learn More
Julabo USA Inc Pair, tube clamps for reinforced tubing 8mm ID or 10mm/12mm ID rubber tubing
SDP
Catalog No. 507534161
Encompass_Preferred
Bath Accessories, Julabo, 2 Tube clamps, size 2, for rubber tubing 10 and 12 mm and Reinforced tubing 8 mm ID
Catalog No. 50-753-4161 Supplier Julabo USA Inc Supplier No. 8970481
Bath Accessories, Julabo, 2 Tube clamps, size 2, for rubber tubing 10 and 12 mm and Reinforced tubing 8 mm ID