Learn More
MSC Vikan Tube Brush, 2 7/16 in Diameter, 19 3/4 in Length, Medium, Pink
Catalog No. 502835527
Encompass
This food-safe tube brush is designed for cleaning the inside of bottles, tubes, and other hard-to-reach spaces, making it ideal for gaps and crevices on machinery and conveyor belts. It features a sturdy polypropylene handle, a stainless steel shaft, and polyester bristles with medium stiffness, effective at removing dirt, grime, and residue.
  • Effectively cleans bottles, tubes, pipes, valves, and hard-to-reach surfaces.
  • Medium bristle stiffness removes dirt, grime, and residue.
  • Flexible, twisted stainless steel shaft.
  • Bristles twisted directly into brush, reducing loss.
  • Durable construction for long-lasting performance.
  • Ergonomic handle for firm, comfortable grip.
  • Pink color for HACCP color-coded system.
  • Certified for EU 10/2011 on food contact, BPA free, dishwasher, and autoclave safe.
Catalog No. 50-283-5527 Supplier MSC Supplier No. 54670377
This food-safe tube brush is designed for cleaning the inside of bottles, tubes, and other hard-to-reach spaces, making it ideal for gaps and crevices on machinery and conveyor belts. It features a sturdy polypropylene handle, a stainless steel shaft, and polyester bristles with medium stiffness, effective at removing dirt, grime, and residue.
  • Effectively cleans bottles, tubes, pipes, valves, and hard-to-reach surfaces.
  • Medium bristle stiffness removes dirt, grime, and residue.
  • Flexible, twisted stainless steel shaft.
  • Bristles twisted directly into brush, reducing loss.
  • Durable construction for long-lasting performance.
  • Ergonomic handle for firm, comfortable grip.
  • Pink color for HACCP color-coded system.
  • Certified for EU 10/2011 on food contact, BPA free, dishwasher, and autoclave safe.