Learn More
MSC Vikan Tube Brush, Ø20 mm, 500 mm, Medium, Pink
Catalog No. 502835949
Encompass
This tube brush is designed for effective cleaning of bottles, tubes, and hard-to-reach surfaces, ideal for critical hygiene and food safety environments. The medium bristles are suitable for scrubbing and dislodging stubborn soils like fatty deposits, greases, and biofilms.
  • Purpose-built for critical hygiene and food safety environments.
  • Medium bristles are ideal for scrubbing stubborn soils.
  • Effective for pipe, tube, and valve cleaning across multiple surface types.
  • Twisted stainless steel shaft provides flexibility for curves and bends.
  • Bristles twisted directly into brush reduce bristle loss.
  • Lightweight design and durable construction.
  • Easy to clean and maintain, with a drop-shaped hanging hole for storage.
Catalog No. 50-283-5949 Supplier MSC Supplier No. 54670328
This tube brush is designed for effective cleaning of bottles, tubes, and hard-to-reach surfaces, ideal for critical hygiene and food safety environments. The medium bristles are suitable for scrubbing and dislodging stubborn soils like fatty deposits, greases, and biofilms.
  • Purpose-built for critical hygiene and food safety environments.
  • Medium bristles are ideal for scrubbing stubborn soils.
  • Effective for pipe, tube, and valve cleaning across multiple surface types.
  • Twisted stainless steel shaft provides flexibility for curves and bends.
  • Bristles twisted directly into brush reduce bristle loss.
  • Lightweight design and durable construction.
  • Easy to clean and maintain, with a drop-shaped hanging hole for storage.