Learn More
MSC Vikan Tube Brush, 0.8" Diameter, 19.7" Length, Medium Stiffness, Purple
Catalog No. 502838249
Encompass
This tube brush effectively cleans bottles, tubes, and hard-to-reach surfaces. It is purpose-built for hygiene and food safety in food manufacturing, retail, and service environments.
  • Hard bristles dislodge stubborn soils like fatty deposits, greases, and biofilms.
  • Effective for pipe, tube, and valve cleaning.
  • Provides effective cleaning across multiple surface types.
  • Twisted stainless steel shaft offers flexibility for curves and bends.
  • Bristles are twisted directly into the brush, reducing bristle loss.
  • Available in multiple diameters for different applications.
  • Effective for tight spaces and crevices.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.
Catalog No. 50-283-8249 Supplier MSC Supplier No. 54670401
This tube brush effectively cleans bottles, tubes, and hard-to-reach surfaces. It is purpose-built for hygiene and food safety in food manufacturing, retail, and service environments.
  • Hard bristles dislodge stubborn soils like fatty deposits, greases, and biofilms.
  • Effective for pipe, tube, and valve cleaning.
  • Provides effective cleaning across multiple surface types.
  • Twisted stainless steel shaft offers flexibility for curves and bends.
  • Bristles are twisted directly into the brush, reducing bristle loss.
  • Available in multiple diameters for different applications.
  • Effective for tight spaces and crevices.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.