Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Tube Closing Plier - Stainless Steel With Durable Insulated Handle

Catalog No. 21125119 Shop All Surgical Design Products
Change view
Click to view available options
Includes:
Type: 1
Type: 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-125-119 Type: 2
21-125-118 Type: 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-125-119 Supplier Surgical Design™ Supplier No. TCP02
Add to Cart
Add to Cart

Tube closing pliers are designed to quickly and securely seal soft metal, plastic, or laminated tubes without leaks.

  • Secure Tube Sealing: Ideal for permanently closing aluminum, lead, or plastic tubes used in labs, manufacturing, or packaging.
  • Flat-Crimp Jaw Design: Creates a tight, uniform seal to prevent leakage and contamination.
  • Stainless Steel Construction: Corrosion-resistant and built for repeated use in industrial and laboratory settings.
  • Comfort Grip Handle: Thick insulated grips provide comfortable, non-slip operation during extended use.
  • Heavy-Duty Riveted Jaws: Ensures proper pressure distribution and long-lasting durability.
  • Multi-Industry Use: Common in pharmaceutical, cosmetic, chemical, and lab environments.

Specifications

Product Type Crimping Pliers
For Use With (Equipment) Ointment Tube
Grade Standard
Handle Material Plastic
Material Stainless Steel
Autoclavable Autoclavable
Includes Type: 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.