Learn More
Andwin Scientific TUBE CLOSURE MULTI PLUG 1000/P
SDP
Catalog No. NC9344939
Encompass_Preferred
The Plug-Type caps are compatible with 12 x 75 mm and 13 x 100 mm culture tubes. These caps feature a disposable, polyethylene construction, an uncommon fluted structure, and snug fit for either plastic or glass tubes. This product is sold in a pack of 1000.
Catalog No. NC9344939 Supplier Andwin Scientific Supplier No. 60819003
The Plug-Type caps are compatible with 12 x 75 mm and 13 x 100 mm culture tubes. These caps feature a disposable, polyethylene construction, an uncommon fluted structure, and snug fit for either plastic or glass tubes. This product is sold in a pack of 1000.