Learn More
Wilmad Labglass TUBE CLR CMPR LOW 100ML
Catalog No. 50807845
Encompass_Preferred
Tube, Color Comparison, Low Form, Unmatched. Graduated at: 100mL; Approximate O.D.: 32mm; Approximate Height: 200mm; Graduated Length: 145-163mm. Tube is graduated at specific points listed.
Catalog No. 50-807-845 Supplier Wilmad Labglass Supplier No. LG10715102
Tube, Color Comparison, Low Form, Unmatched. Graduated at: 100mL; Approximate O.D.: 32mm; Approximate Height: 200mm; Graduated Length: 145-163mm. Tube is graduated at specific points listed.