Learn More
Med Vet International Suction Connector Tubing Medi-Vac 6 Foot Length 0.25 Inch I.D. Sterile Maxi-Grip and Male / Male Connector Clear Smooth OT Surface NonConductive Plastic, N66A, Each
Catalog No. 503044926
Encompass_Preferred
Medi-Vac suction connector tubing helps drain fluids during surgical procedures; drapes easily and springs back to form quickly; avoids kinks and compromised length during use Suction tubing includes maxi-grip connectors; ensure secure attachment to a variety of applications; included male connector enables coupling of additional tubing Clear suction tubing helps maintain visibility of fluids during drainage; thick walls resist collapse when subjected to high-pressure vacuum; tubing is untextured Sterile suction tubing provides safety and hygiene during procedures; intended for single-patient use Tubing is 6 feet in length
Catalog No. 50-304-4926 Supplier Med Vet International Supplier No. 251164EA
Medi-Vac suction connector tubing helps drain fluids during surgical procedures; drapes easily and springs back to form quickly; avoids kinks and compromised length during use Suction tubing includes maxi-grip connectors; ensure secure attachment to a variety of applications; included male connector enables coupling of additional tubing Clear suction tubing helps maintain visibility of fluids during drainage; thick walls resist collapse when subjected to high-pressure vacuum; tubing is untextured Sterile suction tubing provides safety and hygiene during procedures; intended for single-patient use Tubing is 6 feet in length