Learn More
Chemglass Life Sciences Tube, Connecting, "Y" Shaped, CG-300-03 (13.5mm Small Serration, 15.5mm Large Serration) Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG402303
Encompass_Preferred
Tube, Connecting, "Y" Shaped, CG-300-03 (13.5mm Small Serration, 15.5mm Large Serration) Hose Connection. Y shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.
Catalog No. CG402303