Learn More
Med Vet International BD Microtainer Capillary Blood Collection Tube Lithium Heparin Additive 200 µL to 400 µL BD Microgard™ Closure Plastic Tube, 365965, 200/Case
Catalog No. 503044017
Encompass_Preferred
Designed for the collection, transport and processing of skin puncture blood from infants, children, geriatric patients and critical care patients
Catalog No. 50-304-4017 Supplier Med Vet International Supplier No. 450612CS
Designed for the collection, transport and processing of skin puncture blood from infants, children, geriatric patients and critical care patients