Learn More
Med Vet International BD Microtainer Capillary Blood Collection Tube Lithium Heparin Additive 200 µL to 400 µL BD Microgard™ Closure Plastic Tube, 365965, 50/Box
Catalog No. 503045521
Encompass_Preferred
Designed for the collection, transport and processing of skin puncture blood from infants, children, geriatric patients and critical care patients
Catalog No. 50-304-5521 Supplier Med Vet International Supplier No. 450612BX
Designed for the collection, transport and processing of skin puncture blood from infants, children, geriatric patients and critical care patients