Learn More
Chemglass Life Sciences Tube, Culture, N51A, 25 x 100mm
SDP
Catalog No. 50143670
Encompass
Tube, Culture, N51A, 25 x 100mm. The 25 x 100mm culture tube is the preferred culture tube to be used for the AOAC Use-Dilution Method 964.02 and 955.15. The tubes are generally used to hold the disinfectant being tested.
Catalog No. 50-143-670 Supplier Chemglass Life Sciences Supplier No. CLSEPA25100
Tube, Culture, N51A, 25 x 100mm. The 25 x 100mm culture tube is the preferred culture tube to be used for the AOAC Use-Dilution Method 964.02 and 955.15. The tubes are generally used to hold the disinfectant being tested.