Learn More
Waters Corp Waters Corporation Tubing, ETFE, 1/8 in. OD, 0.040 in. ID
Catalog No. 50818897 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing is carefully designed to provide leak-free connections, with low dead volume, and minimal band spreading. For best performance, these components must be clean, and designed to work together. The geometry of the ferrule and the conical port that it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths and diameters are available to provide the flexibility needed when setting up and maintaining a system.
  • Provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Available in a variety of materials, lengths, and diameters
Catalog No. 50-818-897 Supplier Waters Corp Supplier No. WAT088432
This tubing is carefully designed to provide leak-free connections, with low dead volume, and minimal band spreading. For best performance, these components must be clean, and designed to work together. The geometry of the ferrule and the conical port that it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths and diameters are available to provide the flexibility needed when setting up and maintaining a system.
  • Provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Available in a variety of materials, lengths, and diameters