Learn More
Ace Glass, Inc. Tube, distillation, connecting, 24/40 standard taper joints, 0-4 Hi-Vac valve, 125mm center length between joints, No-Air Schlenk
SDP
Catalog No. 50975443
Encompass_Preferred
Tube, distillation, connecting, 24/40 standard taper joints, 0-4 Hi-Vac valve, 125mm center length between joints, No-Air Schlenk
Catalog No. 50-975-443 Supplier Ace Glass, Inc. Supplier No. 781110
Tube, distillation, connecting, 24/40 standard taper joints, 0-4 Hi-Vac valve, 125mm center length between joints, No-Air Schlenk