Learn More
Med Vet International Sheridan Endotracheal Tube, 5.0mm
Catalog No. 502831470
Encompass_Preferred
Sheridan Endotracheal Tube. High quality clear PVC tubes but are inexpensive. Uniform inflatable cuff. Sterile. Endotracheal Tube. 5.0mm. Length 23cm.
Catalog No. 50-283-1470 Supplier Med Vet International Supplier No. JOR835E
Sheridan Endotracheal Tube. High quality clear PVC tubes but are inexpensive. Uniform inflatable cuff. Sterile. Endotracheal Tube. 5.0mm. Length 23cm.