Learn More
New Jersey Fluid Systems LLC TUBE FITTING, M 1/8 X 1/8 M SS
Catalog No. NC2180828
Encompass
Stainless Steel Swagelok Tube Fitting, Male Tube Adapter, 1/8 in. Tube OD x 1/8 in. Male NPT
This item is not returnable. View return policy
Catalog No. NC2180828 Supplier New Jersey Fluid Systems LLC Supplier No. SS2TA12
Stainless Steel Swagelok Tube Fitting, Male Tube Adapter, 1/8 in. Tube OD x 1/8 in. Male NPT