Learn More
New Jersey Fluid Systems LLC Swagelok HC-810-6 Tube Fitting Union, 1/2 In, Superior Corrosion Resistance, Gas-Tight Seal
Catalog No. NC3714877
Encompass
The Swagelok HC-810-6 is a 1/2 in tube fitting union designed for leak-tight connections in high-pressure systems. Its robust Alloy C-276 construction ensures superior corrosion resistance, making it ideal for industrial applications like chemical processing and oil & gas. The two-ferrule design offers reliable gas-tight seals with easy installation and reusability, setting it apart from competitors by maintaining system integrity and minimizing downtime.
Catalog No. NC3714877 Supplier New Jersey Fluid Systems LLC Supplier No. HC8106
The Swagelok HC-810-6 is a 1/2 in tube fitting union designed for leak-tight connections in high-pressure systems. Its robust Alloy C-276 construction ensures superior corrosion resistance, making it ideal for industrial applications like chemical processing and oil & gas. The two-ferrule design offers reliable gas-tight seals with easy installation and reusability, setting it apart from competitors by maintaining system integrity and minimizing downtime.