Learn More
Research Products International Corp Tube Foam Inserts for Vortex Genie, 9-16mm, 2/pk
SDP
Catalog No. 501978462
Encompass
Two high-density Foam Inserts to accommodate 9 - 16 mm diameter tubes. The insert fits into the Recessed Platform for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Catalog No. 50-197-8462 Supplier Research Products International Corp Supplier No. 155318
Two high-density Foam Inserts to accommodate 9 - 16 mm diameter tubes. The insert fits into the Recessed Platform for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.