Learn More
Research Products International Corp Tube Freezer Boxes for 1.5-2.0ml Tubes, 5 1/2 x 5 1/2 x 2 Inches, 4 per Case
SDP
Catalog No. 501367678
Encompass
Polypropylene Tube Freezer Boxes are ideal for storing mirco-tubes or cryogenic vials in freezers. Durable boxes conveniently transport samples and will not break down or decompose like cardboard boxes. Boxes feature alpha numeric grid for easy identification and withstand temperatures down to -80°C.
Supplied with lid and sold 4 racks per case.
Catalog No. 50-136-7678 Supplier Research Products International Corp Supplier No. 181052
Polypropylene Tube Freezer Boxes are ideal for storing mirco-tubes or cryogenic vials in freezers. Durable boxes conveniently transport samples and will not break down or decompose like cardboard boxes. Boxes feature alpha numeric grid for easy identification and withstand temperatures down to -80°C.
Supplied with lid and sold 4 racks per case.