Learn More
Wilmad Labglass TUBE GAS COLL PTFE S/C 125ML
Supplier:  Wilmad Labglass LG8601T104
Encompass_Preferred
Gas Collecting Tube, with PTFE Plugs. Capacity: 125mL; Body OD: 38mm; Body Length: 300mm; Stopcock Size: 2mm. Unit has a side outlet in body For a self sealing syringe stopper. Useful in gas chromatography or mass spectroscopy.
Catalog No. 50-805-608