Learn More
Wilmad Labglass TUBE GAS DISP CYL 12 C
Catalog No. 50803087
Encompass_Preferred
Gas Dispersion Tube, Cylindrical. Porosity: Coarse; Fritted Glass Cylinder OD: 12mm; Fritted Glass Cylinder Length: 25mm; Tubing OD: 8mm; Approximate Overall Length: 250mm.
Catalog No. 50-803-087 Supplier Wilmad Labglass Supplier No. LG8680110
Gas Dispersion Tube, Cylindrical. Porosity: Coarse; Fritted Glass Cylinder OD: 12mm; Fritted Glass Cylinder Length: 25mm; Tubing OD: 8mm; Approximate Overall Length: 250mm.