Learn More
Chemglass Life Sciences Gas Dispersion Tube, 30mm Diameter Coarse Frit
SDP
Catalog No. 50873742
Encompass_Preferred
Gas Dispersion Tube, 30mm Diameter Coarse Frit. Fritted gas dispersion tube with tubing sealed on one end.
Catalog No. 50-873-742 Supplier Chemglass Life Sciences Supplier No. CG20411
Gas Dispersion Tube, 30mm Diameter Coarse Frit. Fritted gas dispersion tube with tubing sealed on one end.