Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 20mm Diameter, Fine Porosity, Tube O.D./25mm
SDP
Catalog No. 501531401
Encompass_Preferred
Glassblowers Fritted Tube, Straight, Frit 20mm Diameter, Fine Porosity, Tube O.D./25mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.
Catalog No. 50-153-1401 Supplier Chemglass Life Sciences Supplier No. CG20205
Glassblowers Fritted Tube, Straight, Frit 20mm Diameter, Fine Porosity, Tube O.D./25mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.