Learn More
Andwin Scientific TUBE GRAD NAT 0.5ML 1K/PK
SDP
Catalog No. NC0910936
Encompass_Preferred
The disposable 0.5 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in a pack of 1000 natural-colored tubes.
Catalog No. NC0910936 Supplier Andwin Scientific Supplier No. 89000010
The disposable 0.5 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in a pack of 1000 natural-colored tubes.