Learn More
Research Products International Corp Vortex-Genie 2 Tube Holder, Horizontal, 15ml Tubes, Holds 12 Tubes
SDP
Catalog No. 50998334
Encompass
Tube Holder holds twelve 15 ml tubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-998-334 Supplier Research Products International Corp Supplier No. 155586
Tube Holder holds twelve 15 ml tubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.