Learn More
Research Products International Corp Vortex-Genie 2 Tube Holder, Horizontal, 1.5-2.0ml Tubes, Holds 24 Tubes
SDP
Catalog No. 50998333
Encompass
Tube Holder holds twenty-four microtubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-998-333 Supplier Research Products International Corp Supplier No. 155585
Tube Holder holds twenty-four microtubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.